Product Details

SNP ID
rs114666749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89241102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAAGAACTGACCTTACCTGTGA[C/T]TAATGGCATGGTTACTTAGCCCTGG
Phenotype
MIM: 611241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPRIN3 PubMed Links

Gene Details

Gene
GPRIN3
Gene Name
GPRIN family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198281.2 9326 Intron NP_938022.2
XM_005262936.3 9326 UTR 3 XP_005262993.1
XM_005262937.4 9326 UTR 3 XP_005262994.1
XM_017008043.1 9326 UTR 3 XP_016863532.1
XM_017008044.1 9326 UTR 3 XP_016863533.1

View Full Product Details