Product Details

SNP ID
rs116076523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99879648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCATGTGTTACGTAACAATGTG[A/G]ATACATTTCGAGACATGCTTTGTTA
Phenotype
MIM: 605768 MIM: 603296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAPP1 PubMed Links

Gene Details

Gene
DAPP1
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
There are no transcripts associated with this gene.

Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243736.1 2970 UTR 3 NP_001230665.1
NM_021970.3 2970 UTR 3 NP_068805.1

View Full Product Details