Product Details

SNP ID
rs113542593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99885520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGACAACTGAAATCAGCAAAT[A/C]ATTTTATAATTATGAACTTACCTGG
Phenotype
MIM: 603296
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LAMTOR3 PubMed Links

Gene Details

Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243736.1 Intron NP_001230665.1
NM_021970.3 Intron NP_068805.1

View Full Product Details