Product Details

SNP ID
rs116393199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77876460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTCTACCCCAAGTTCTATTCC[A/C]GTATGGTTTTAAATGTGCTTTTCAA
Phenotype
MIM: 611821
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPL1 PubMed Links

Gene Details

Gene
MRPL1
Gene Name
mitochondrial ribosomal protein L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020236.3 Intron NP_064621.3
XM_011532198.2 Intron XP_011530500.1
XM_011532199.1 Intron XP_011530501.1

View Full Product Details