Product Details

SNP ID
rs115815850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139717919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGCGTCAATTGCAGAGAACAA[C/T]TGTACAGGACTTTGGAAGAGGACTA
Phenotype
MIM: 608991 MIM: 601733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAML3 PubMed Links

Gene Details

Gene
MAML3
Gene Name
mastermind like transcriptional coactivator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018717.4 5558 UTR 3 NP_061187.2
Gene
MGST2
Gene Name
microsomal glutathione S-transferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204366.1 5558 Intron NP_001191295.1
NM_001204367.1 5558 Intron NP_001191296.1
NM_001204368.1 5558 Intron NP_001191297.1
NM_002413.4 5558 Intron NP_002404.1
XM_006714221.2 5558 Intron XP_006714284.1
XM_011531972.2 5558 Intron XP_011530274.1
XM_017008211.1 5558 Intron XP_016863700.1
XM_017008212.1 5558 Intron XP_016863701.1
XM_017008213.1 5558 Intron XP_016863702.1
XM_017008214.1 5558 Intron XP_016863703.1

View Full Product Details