Product Details
- SNP ID
-
rs116164074
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:151278079 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGTATGCTGAGAGAGCAAAAAA[C/T]TTAACAGGTAGCAAAGAAGAAAGAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS48
PubMed Links
Gene Details
- Gene
- PRSS48
- Gene Name
- protease, serine 48
View Full Product Details