Product Details
- SNP ID
-
rs113646668
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:108247224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAAGATTGAGAAAAACGCTCCCAC[A/G]ATGGAAAAAAAGCCAGAACTGTTTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SULT1C3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9308806,rs9646947] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SULT1C3
- Gene Name
- sulfotransferase family 1C member 3
View Full Product Details