Product Details
- SNP ID
-
rs115471979
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:238923082 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCACACACACTCACTTTTGAGCA[C/T]ATCCTCTGTGGAGACTTGGAGCTAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ43879
PubMed Links
Gene Details
- Gene
- FLJ43879
- Gene Name
- FLJ43879 protein
There are no transcripts associated with this gene.
View Full Product Details