Product Details

SNP ID
rs115424252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24014557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCATTCAAAATAATGATCTAT[A/G]CCAGTTGACTTAGAGTGATTTCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD2B PubMed Links

Gene Details

Gene
MFSD2B
Gene Name
major facilitator superfamily domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080473.1 Intron NP_001073942.1
XM_005264301.4 Intron XP_005264358.1
XM_017004063.1 Intron XP_016859552.1
XM_017004064.1 Intron XP_016859553.1
XM_017004065.1 Intron XP_016859554.1
XM_017004066.1 Intron XP_016859555.1
XM_017004067.1 Intron XP_016859556.1
XM_017004068.1 Intron XP_016859557.1
XM_017004069.1 Intron XP_016859558.1
XM_017004070.1 Intron XP_016859559.1

View Full Product Details