Product Details

SNP ID
rs113961704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24036698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAGTGAATGTCAATCAACAAGG[A/G]AATGGCTAAATAAAGAGCATACTCT
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322963.1 Intron NP_001309892.1
NM_001322964.1 Intron NP_001309893.1
NM_004116.4 Intron NP_004107.1
NM_054033.3 Intron NP_473374.1
XM_011532699.2 Intron XP_011531001.1
XM_017003593.1 Intron XP_016859082.1
XM_017003594.1 Intron XP_016859083.1
XM_017003595.1 Intron XP_016859084.1
XM_017003596.1 Intron XP_016859085.1
Gene
MFSD2B
Gene Name
major facilitator superfamily domain containing 2B
There are no transcripts associated with this gene.

Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 Intron NP_001135791.1
NM_025203.2 Intron NP_079479.1
XM_017005029.1 Intron XP_016860518.1

View Full Product Details