Product Details

SNP ID
rs117386259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55866825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCAAGTTTCACCAGATAGTGTA[G/T]ACTTAGCTCTCTTGAAGAAGACCAG
Phenotype
MIM: 601548
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFEMP1 PubMed Links

Gene Details

Gene
EFEMP1
Gene Name
EGF containing fibulin like extracellular matrix protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039348.2 2190 UTR 3 NP_001034437.1
NM_001039349.2 2190 UTR 3 NP_001034438.1
XM_005264205.4 2190 UTR 3 XP_005264262.1
XM_017003586.1 2190 UTR 3 XP_016859075.1

View Full Product Details