Product Details

SNP ID
rs112070345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61069500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGCGAATCTCTGCTTTGTACTC[C/T]CGTCCTAAAATAGATCATTTTTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1841 PubMed Links

Gene Details

Gene
KIAA1841
Gene Name
KIAA1841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264605.3 Intron XP_005264662.1
XM_005264606.4 Intron XP_005264663.1
XM_005264607.2 Intron XP_005264664.1
XM_011533126.2 Intron XP_011531428.1
XM_011533127.2 Intron XP_011531429.1
XM_011533128.1 Intron XP_011531430.1
XM_011533129.2 Intron XP_011531431.1
XM_011533131.2 Intron XP_011531433.1
XM_017005108.1 Intron XP_016860597.1
XM_017005109.1 Intron XP_016860598.1

View Full Product Details