Product Details

SNP ID
rs112148438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25383649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAATGAGGAAGCTCTTGGTGAT[C/T]GACTGACCTTGTCACCTGGAAGGTA
Phenotype
MIM: 602415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DTNB PubMed Links

Gene Details

Gene
DTNB
Gene Name
dystrobrevin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256303.1 Intron NP_001243232.1
NM_001256304.1 Intron NP_001243233.1
NM_001256308.2 Intron NP_001243237.1
NM_001320932.1 Intron NP_001307861.1
NM_001320933.1 Intron NP_001307862.1
NM_001320934.1 Intron NP_001307863.1
NM_001320935.1 Intron NP_001307864.1
NM_001320936.1 Intron NP_001307865.1
NM_001320937.1 Intron NP_001307866.1
NM_021907.4 Intron NP_068707.1
NM_033147.3 Intron NP_149159.2
NM_033148.3 Intron NP_149160.1
NM_183360.2 Intron NP_899204.1
NM_183361.2 Intron NP_899205.1
XM_005264182.1 Intron XP_005264239.1
XM_011532671.1 Intron XP_011530973.1
XM_011532672.2 Intron XP_011530974.1
XM_011532673.1 Intron XP_011530975.1
XM_011532674.1 Intron XP_011530976.1
XM_011532675.1 Intron XP_011530977.1
XM_011532676.1 Intron XP_011530978.1
XM_011532677.2 Intron XP_011530979.1
XM_011532679.1 Intron XP_011530981.1
XM_011532680.2 Intron XP_011530982.1
XM_011532681.2 Intron XP_011530983.1
XM_017003529.1 Intron XP_016859018.1
XM_017003530.1 Intron XP_016859019.1
XM_017003531.1 Intron XP_016859020.1
XM_017003532.1 Intron XP_016859021.1
XM_017003533.1 Intron XP_016859022.1
XM_017003534.1 Intron XP_016859023.1
XM_017003535.1 Intron XP_016859024.1
XM_017003536.1 Intron XP_016859025.1
XM_017003537.1 Intron XP_016859026.1
XM_017003538.1 Intron XP_016859027.1
XM_017003539.1 Intron XP_016859028.1
XM_017003540.1 Intron XP_016859029.1
XM_017003541.1 Intron XP_016859030.1
XM_017003542.1 Intron XP_016859031.1
XM_017003543.1 Intron XP_016859032.1
XM_017003544.1 Intron XP_016859033.1
XM_017003545.1 Intron XP_016859034.1

View Full Product Details