Product Details

SNP ID
rs112465352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73229495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGAGCAATCTGGTCCTGGATGA[A/G]GAAACCGTTGCTGAGTTCCCCGCAG
Phenotype
MIM: 605140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT7 PubMed Links

Gene Details

Gene
CCT7
Gene Name
chaperonin containing TCP1 subunit 7
There are no transcripts associated with this gene.

Gene
PRADC1
Gene Name
protease associated domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032319.1 312 Missense Mutation CTC,TTC L82F NP_115695.1
Gene
SMYD5
Gene Name
SMYD family member 5
There are no transcripts associated with this gene.

View Full Product Details