Product Details

SNP ID
rs114786974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26863412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCTGGAGTTTCCATTCCTTTC[C/T]TATCACTGCTTCTCAAATCTTGCCC
Phenotype
MIM: 608383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYSL5 PubMed Links

Gene Details

Gene
DPYSL5
Gene Name
dihydropyrimidinase like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253723.1 Intron NP_001240652.1
NM_001253724.1 Intron NP_001240653.1
NM_020134.3 Intron NP_064519.2

View Full Product Details