Product Details
- SNP ID
-
rs113622499
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:27256466 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTCATGGGTTGCCCGGACTCCT[A/G]GCACCGACAACAGCGTATCCCGCAC
- Phenotype
-
MIM: 602878
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC30A3
PubMed Links
Gene Details
- Gene
- SLC30A3
- Gene Name
- solute carrier family 30 member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318949.1 |
981 |
Missense Mutation |
CCA,CTA |
P308L |
NP_001305878.1 |
NM_001318950.1 |
981 |
Missense Mutation |
CCA,CTA |
P300L |
NP_001305879.1 |
NM_001318951.1 |
981 |
Missense Mutation |
CCA,CTA |
P300L |
NP_001305880.1 |
NM_003459.4 |
981 |
Missense Mutation |
CCA,CTA |
P313L |
NP_003450.2 |
XM_005264547.1 |
981 |
Nonsense Mutation |
CAG,TAG |
Q262* |
XP_005264604.1 |
XM_005264548.3 |
981 |
Nonsense Mutation |
CAG,TAG |
Q257* |
XP_005264605.1 |
XM_006712100.2 |
981 |
Nonsense Mutation |
CAG,TAG |
Q213* |
XP_006712163.1 |
XM_011533102.2 |
981 |
Missense Mutation |
CCA,CTA |
P240L |
XP_011531404.1 |
XM_011533103.2 |
981 |
Missense Mutation |
CCA,CTA |
P199L |
XP_011531405.1 |
XM_017004873.1 |
981 |
Nonsense Mutation |
CAG,TAG |
Q249* |
XP_016860362.1 |
XM_017004874.1 |
981 |
Nonsense Mutation |
CAG,TAG |
Q249* |
XP_016860363.1 |
XM_017004875.1 |
981 |
Nonsense Mutation |
CAG,TAG |
Q240* |
XP_016860364.1 |
View Full Product Details