Product Details
- SNP ID
-
rs112667682
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:27447858 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATCAGCCCAGCTATGATAGGCC[C/T]CGGCACAGTTGGTTCCAGGAGAGCT
- Phenotype
-
MIM: 607386
MIM: 606010
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFT172
PubMed Links
Gene Details
- Gene
- IFT172
- Gene Name
- intraflagellar transport 172
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015662.2 |
4677 |
Missense Mutation |
GAG,GGG |
E1498G |
NP_056477.1 |
XM_005264254.1 |
4677 |
Missense Mutation |
GAG,GGG |
E1476G |
XP_005264311.1 |
XM_006711986.3 |
4677 |
Missense Mutation |
GAG,GGG |
E1477G |
XP_006712049.1 |
XM_006711987.1 |
4677 |
Missense Mutation |
GAG,GGG |
E1498G |
XP_006712050.1 |
XM_011532757.2 |
4677 |
Missense Mutation |
GAG,GGG |
E1271G |
XP_011531059.1 |
XM_011532758.1 |
4677 |
Intron |
|
|
XP_011531060.1 |
XM_011532759.2 |
4677 |
Missense Mutation |
GAG,GGG |
E978G |
XP_011531061.1 |
XM_011532760.2 |
4677 |
Missense Mutation |
GAG,GGG |
E853G |
XP_011531062.1 |
XM_017003790.1 |
4677 |
Missense Mutation |
GAG,GGG |
E1455G |
XP_016859279.1 |
XM_017003791.1 |
4677 |
Missense Mutation |
GAG,GGG |
E1271G |
XP_016859280.1 |
XM_017003792.1 |
4677 |
Intron |
|
|
XP_016859281.1 |
XM_017003793.1 |
4677 |
Missense Mutation |
GAG,GGG |
E877G |
XP_016859282.1 |
XM_017003794.1 |
4677 |
Missense Mutation |
GAG,GGG |
E877G |
XP_016859283.1 |
XM_017003795.1 |
4677 |
Missense Mutation |
GAG,GGG |
E809G |
XP_016859284.1 |
- Gene
- KRTCAP3
- Gene Name
- keratinocyte associated protein 3
There are no transcripts associated with this gene.
- Gene
- NRBP1
- Gene Name
- nuclear receptor binding protein 1
There are no transcripts associated with this gene.
View Full Product Details