Product Details

SNP ID
rs112654860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86604223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTCCTGGAGAATTGTGAAGTA[C/T]GCTTTGCTTATCTTCAAATACTTCC
Phenotype
MIM: 602507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105374845 PubMed Links

Gene Details

Gene
LOC105374845
Gene Name
uncharacterized LOC105374845
There are no transcripts associated with this gene.

Gene
RNF103
Gene Name
ring finger protein 103
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198951.1 1681 Missense Mutation ATA,GTA I556V NP_001185880.1
NM_001198952.1 1681 Intron NP_001185881.1
NM_005667.3 1681 Missense Mutation ATA,GTA I560V NP_005658.1
Gene
RNF103-CHMP3
Gene Name
RNF103-CHMP3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198954.1 1681 Intron NP_001185883.1

View Full Product Details