Product Details

SNP ID
rs114302776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28900719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCAACAAGTTAAATGAGACCAT[C/T]AGACTTTGTACTTTGGCTTTTAATT
Phenotype
MIM: 616195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR43 PubMed Links

Gene Details

Gene
WDR43
Gene Name
WD repeat domain 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015131.2 Intron NP_055946.1

View Full Product Details