Product Details

SNP ID
rs112237586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69024055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCTTAGGTGAAGAATACTTAC[C/T]CTTAGCTGAGTTCTAAGTTTGGGTA
Phenotype
MIM: 601126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMF1 PubMed Links

Gene Details

Gene
TMF1
Gene Name
TATA element modulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007114.2 3385 Silent Mutation AGA,AGG R1046R NP_009045.2
XM_011534056.2 3385 Silent Mutation AGA,AGG R1049R XP_011532358.1

View Full Product Details