Product Details

SNP ID
rs112814470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:71681701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGGCTAGCCCTACGTCTTTAA[A/C]AAGGGACAAGTTCGGCCAAGGCTCA
Phenotype
MIM: 609896
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EIF4E3 PubMed Links

Gene Details

Gene
EIF4E3
Gene Name
eukaryotic translation initiation factor 4E family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134649.2 3797 UTR 3 NP_001128121.1
NM_001134650.1 3797 UTR 3 NP_001128122.1
NM_001134651.1 3797 UTR 3 NP_001128123.1
NM_001282886.1 3797 UTR 3 NP_001269815.1
NM_173359.4 3797 UTR 3 NP_775495.1
XM_011533651.2 3797 UTR 3 XP_011531953.1
XM_017006278.1 3797 UTR 3 XP_016861767.1
XM_017006279.1 3797 UTR 3 XP_016861768.1
XM_017006280.1 3797 UTR 3 XP_016861769.1
XM_017006281.1 3797 UTR 3 XP_016861770.1
XM_017006282.1 3797 UTR 3 XP_016861771.1

View Full Product Details