Product Details

SNP ID
rs111477481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15398189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTCAGTTGAACTCTCCACTGCA[A/G]TGCTGAGATTTTTTTTTTTTTTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL6 PubMed Links

Gene Details

Gene
METTL6
Gene Name
methyltransferase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301790.1 Intron NP_001288719.1
NM_001301791.1 Intron NP_001288720.1
NM_001301792.1 Intron NP_001288721.1
NM_152396.3 Intron NP_689609.2
XM_005264867.3 Intron XP_005264924.1
XM_005264870.4 Intron XP_005264927.1
XM_006712970.3 Intron XP_006713033.1
XM_006712972.3 Intron XP_006713035.1
XM_011533357.2 Intron XP_011531659.1
XM_017005718.1 Intron XP_016861207.1
XM_017005719.1 Intron XP_016861208.1
XM_017005720.1 Intron XP_016861209.1
XM_017005721.1 Intron XP_016861210.1
XM_017005722.1 Intron XP_016861211.1
XM_017005723.1 Intron XP_016861212.1
XM_017005724.1 Intron XP_016861213.1

View Full Product Details