Product Details

SNP ID
rs112075281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:20042066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCCCAGTGCTCCTCAAACTTTA[A/G]TGTGCATGGGAATCTCCTGGGGAAG
Phenotype
MIM: 602303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAT2B PubMed Links

Gene Details

Gene
KAT2B
Gene Name
lysine acetyltransferase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003884.4 Intron NP_003875.3
XM_005265528.4 Intron XP_005265585.1
XM_017007423.1 Intron XP_016862912.1
XM_017007424.1 Intron XP_016862913.1

View Full Product Details