Product Details

SNP ID
rs115153721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24121600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGGTTGGGCAGGAAAGTTAAG[G/T]GTGCACATGGTCTGCAGGTGTTTTC
Phenotype
MIM: 190160
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 3092 UTR 3 NP_000452.2
NM_001128176.2 3092 UTR 3 NP_001121648.1
NM_001128177.1 3092 UTR 3 NP_001121649.1
NM_001252634.1 3092 UTR 3 NP_001239563.1
XM_005265421.4 3092 UTR 3 XP_005265478.1
XM_005265423.4 3092 UTR 3 XP_005265480.1
XM_005265424.3 3092 UTR 3 XP_005265481.1
XM_006713317.3 3092 UTR 3 XP_006713380.1
XM_006713318.3 3092 UTR 3 XP_006713381.1
XM_011534046.2 3092 UTR 3 XP_011532348.1
XM_011534047.2 3092 UTR 3 XP_011532349.1
XM_011534048.2 3092 UTR 3 XP_011532350.1
XM_011534049.2 3092 UTR 3 XP_011532351.1
XM_011534050.2 3092 UTR 3 XP_011532352.1
XM_011534051.2 3092 UTR 3 XP_011532353.1
XM_011534052.2 3092 UTR 3 XP_011532354.1
XM_011534053.2 3092 UTR 3 XP_011532355.1
XM_017007108.1 3092 UTR 3 XP_016862597.1
XM_017007109.1 3092 UTR 3 XP_016862598.1
XM_017007110.1 3092 UTR 3 XP_016862599.1
XM_017007111.1 3092 UTR 3 XP_016862600.1
XM_017007112.1 3092 UTR 3 XP_016862601.1
XM_017007113.1 3092 UTR 3 XP_016862602.1
XM_017007114.1 3092 UTR 3 XP_016862603.1
XM_017007115.1 3092 UTR 3 XP_016862604.1

View Full Product Details