Product Details

SNP ID
rs114555929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39097848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATGACAGAGGACTCAGTGTTT[A/G]GGACTTGAAAGGGGGTGGTCCCAGG
Phenotype
MIM: 606867 MIM: 611430 MIM: 612167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GORASP1 PubMed Links

Gene Details

Gene
GORASP1
Gene Name
golgi reassembly stacking protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278789.1 2453 UTR 3 NP_001265718.1
NM_001278790.1 2453 UTR 3 NP_001265719.1
NM_031899.3 2453 UTR 3 NP_114105.1
XM_006713301.3 2453 UTR 3 XP_006713364.1
XM_011534020.2 2453 UTR 3 XP_011532322.1
XM_011534021.2 2453 UTR 3 XP_011532323.1
XM_017007050.1 2453 UTR 3 XP_016862539.1
XM_017007051.1 2453 UTR 3 XP_016862540.1
XM_017007052.1 2453 UTR 3 XP_016862541.1
Gene
TTC21A
Gene Name
tetratricopeptide repeat domain 21A
There are no transcripts associated with this gene.

Gene
WDR48
Gene Name
WD repeat domain 48
There are no transcripts associated with this gene.

View Full Product Details