Product Details

SNP ID
rs114668300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39141917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACAAGGTGTTCCCACAAGCACA[C/T]AGGCTGGCAAGGCCTCCTGGGCAAG
Phenotype
MIM: 606458 MIM: 611430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSRNP1 PubMed Links

Gene Details

Gene
CSRNP1
Gene Name
cysteine and serine rich nuclear protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320559.1 3122 UTR 3 NP_001307488.1
NM_001320560.1 3122 UTR 3 NP_001307489.1
NM_033027.3 3122 UTR 3 NP_149016.2
XM_017007048.1 3122 UTR 3 XP_016862537.1
XM_017007049.1 3122 UTR 3 XP_016862538.1
Gene
MIR6822
Gene Name
microRNA 6822
There are no transcripts associated with this gene.

Gene
TTC21A
Gene Name
tetratricopeptide repeat domain 21A
There are no transcripts associated with this gene.

View Full Product Details