Product Details

SNP ID
rs116466773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39816988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGCCCACTTATATGTAGGTCCC[C/T]ATTGTGATGTCTTAAGATAGCACTA
Phenotype
MIM: 611790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYRIP PubMed Links

Gene Details

Gene
MYRIP
Gene Name
myosin VIIA and Rab interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284423.1 Intron NP_001271352.1
NM_001284424.1 Intron NP_001271353.1
NM_001284425.1 Intron NP_001271354.1
NM_001284426.1 Intron NP_001271355.1
NM_015460.3 Intron NP_056275.2
XM_011533575.1 Intron XP_011531877.1

View Full Product Details