Product Details

SNP ID
rs112257688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46498170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGACAAGGGCCTTCTTCCCAA[C/T]GTCCTGAAGCCAGGCTGGATGCAAT
Phenotype
MIM: 607181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTP3 PubMed Links

Gene Details

Gene
RTP3
Gene Name
receptor transporter protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031440.1 176 Silent Mutation AAC,AAT N36N NP_113628.1

View Full Product Details