Product Details

SNP ID
rs115044799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47011645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTGGGAGGTACCCCAAGTGGG[G/T]AGGTCACACCCAAGTCCAGCCACAG
Phenotype
MIM: 614169 MIM: 612778
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NBEAL2 PubMed Links

Gene Details

Gene
NBEAL2
Gene Name
neurobeachin like 2
There are no transcripts associated with this gene.

Gene
NRADDP
Gene Name
neurotrophin receptor associated death domain, pseudogene
There are no transcripts associated with this gene.

Gene
SETD2
Gene Name
SET domain containing 2
There are no transcripts associated with this gene.

View Full Product Details