Product Details

SNP ID
rs117583613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48561890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGCCCAGGACAGTCCGTGCGT[A/G]AAGTGGGACAGAGTTTGGTTGGAGC
Phenotype
MIM: 120120 MIM: 605320 MIM: 605902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL7A1 PubMed Links

Gene Details

Gene
COL7A1
Gene Name
collagen type VII alpha 1
There are no transcripts associated with this gene.

Gene
PFKFB4
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317134.1 494 Intron NP_001304063.1
NM_001317135.1 494 Intron NP_001304064.1
NM_001317136.1 494 Intron NP_001304065.1
NM_001317137.1 494 Intron NP_001304066.1
NM_001317138.1 494 Intron NP_001304067.1
NM_004567.3 494 Intron NP_004558.1
XM_011533829.2 494 UTR 5 XP_011532131.1
XM_017006614.1 494 Intron XP_016862103.1
XM_017006615.1 494 Intron XP_016862104.1
XM_017006616.1 494 Intron XP_016862105.1
XM_017006617.1 494 Intron XP_016862106.1
Gene
UCN2
Gene Name
urocortin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033199.3 494 UTR 3 NP_149976.1

View Full Product Details