Product Details
- SNP ID
-
rs111339760
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:49110961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCTGTTCTCACCAGGCCTCCTC[A/G]GGTGCCAGCACCTCAGGCCGTGTGA
- Phenotype
-
MIM: 603727
MIM: 614471
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
QARS
PubMed Links
Gene Details
- Gene
- QARS
- Gene Name
- glutaminyl-tRNA synthetase
There are no transcripts associated with this gene.
- Gene
- USP19
- Gene Name
- ubiquitin specific peptidase 19
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199160.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1176P |
NP_001186089.1 |
NM_001199161.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1178P |
NP_001186090.1 |
NM_001199162.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1166P |
NP_001186091.1 |
NM_006677.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1075P |
NP_006668.1 |
XM_005264823.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1180P |
XP_005264880.1 |
XM_005264824.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1180P |
XP_005264881.1 |
XM_005264825.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1178P |
XP_005264882.1 |
XM_005264826.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1178P |
XP_005264883.1 |
XM_005264827.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1165P |
XP_005264884.1 |
XM_005264829.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1180P |
XP_005264886.1 |
XM_005264830.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1079P |
XP_005264887.1 |
XM_005264831.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1062P |
XP_005264888.1 |
XM_006712946.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1179P |
XP_006713009.1 |
XM_006712947.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1162P |
XP_006713010.1 |
XM_006712948.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1176P |
XP_006713011.1 |
XM_006712949.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1130P |
XP_006713012.1 |
XM_006712950.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1163P |
XP_006713013.1 |
XM_006712951.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1162P |
XP_006713014.1 |
XM_006712952.2 |
3768 |
Silent Mutation |
CCC,CCT |
P1161P |
XP_006713015.1 |
XM_006712953.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1161P |
XP_006713016.1 |
XM_017005615.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1177P |
XP_016861104.1 |
XM_017005616.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1175P |
XP_016861105.1 |
XM_017005617.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1163P |
XP_016861106.1 |
XM_017005618.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1178P |
XP_016861107.1 |
XM_017005619.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1177P |
XP_016861108.1 |
XM_017005620.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1175P |
XP_016861109.1 |
XM_017005621.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1128P |
XP_016861110.1 |
XM_017005622.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1165P |
XP_016861111.1 |
XM_017005623.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1126P |
XP_016861112.1 |
XM_017005624.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1126P |
XP_016861113.1 |
XM_017005625.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1077P |
XP_016861114.1 |
XM_017005626.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1075P |
XP_016861115.1 |
XM_017005627.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1064P |
XP_016861116.1 |
XM_017005628.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1062P |
XP_016861117.1 |
XM_017005629.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1060P |
XP_016861118.1 |
XM_017005630.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1079P |
XP_016861119.1 |
XM_017005631.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1077P |
XP_016861120.1 |
XM_017005632.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1077P |
XP_016861121.1 |
XM_017005633.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1075P |
XP_016861122.1 |
XM_017005634.1 |
3768 |
Silent Mutation |
CCC,CCT |
P1064P |
XP_016861123.1 |
XM_017005635.1 |
3768 |
Silent Mutation |
CCC,CCT |
P628P |
XP_016861124.1 |
View Full Product Details