Product Details

SNP ID
rs114730569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51971622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTCTCGGAAGAAGAGGGCCT[A/G]GCCCTGCACCTGGATGGTGCCCTCG
Phenotype
MIM: 608503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD14A PubMed Links

Gene Details

Gene
ABHD14A
Gene Name
abhydrolase domain containing 14A
There are no transcripts associated with this gene.

Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
There are no transcripts associated with this gene.

Gene
ABHD14B
Gene Name
abhydrolase domain containing 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146314.1 743 Nonsense Mutation CAG,TAG Q17* NP_001139786.1
NM_001254753.1 743 Intron NP_001241682.1
NM_032750.2 743 Nonsense Mutation CAG,TAG Q17* NP_116139.1
Gene
PCBP4
Gene Name
poly(rC) binding protein 4
There are no transcripts associated with this gene.

View Full Product Details