Product Details

SNP ID
rs114535630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52688913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGAAATGATGAGGGTCAACATT[A/C]TTCATACCAAAGTGAAGACATGAGA
Phenotype
MIM: 608011 MIM: 606083
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLT8D1 PubMed Links

Gene Details

Gene
GLT8D1
Gene Name
glycosyltransferase 8 domain containing 1
There are no transcripts associated with this gene.

Gene
GNL3
Gene Name
G protein nucleolar 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014366.4 Intron NP_055181.3
NM_206825.1 Intron NP_996561.1
NM_206826.1 Intron NP_996562.1
Gene
PBRM1
Gene Name
polybromo 1
There are no transcripts associated with this gene.

Gene
SNORD136
Gene Name
small nucleolar RNA, C/D box 136
There are no transcripts associated with this gene.

Gene
SNORD19
Gene Name
small nucleolar RNA, C/D box 19
There are no transcripts associated with this gene.

Gene
SNORD19B
Gene Name
small nucleolar RNA, C/D box 19B
There are no transcripts associated with this gene.

Gene
SNORD69
Gene Name
small nucleolar RNA, C/D box 69
There are no transcripts associated with this gene.

View Full Product Details