Product Details

SNP ID
rs112485971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53848672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTCTCTCCCACAGACCGTTCA[A/G]TGTGGCTCTGAAACTGGTAGGTGCA
Phenotype
MIM: 605458
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHDH PubMed Links

Gene Details

Gene
CHDH
Gene Name
choline dehydrogenase
There are no transcripts associated with this gene.

Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 105 Silent Mutation CAA,CAG Q23Q NP_061195.2
XM_005265310.4 105 Silent Mutation CAA,CAG Q23Q XP_005265367.1
XM_005265311.4 105 Missense Mutation CAA,CAG Q23Q XP_005265368.1
XM_005265312.4 105 Silent Mutation CAA,CAG Q23Q XP_005265369.1
XM_011533941.2 105 Silent Mutation CAA,CAG Q23Q XP_011532243.1
XM_017006804.1 105 UTR 5 XP_016862293.1
XM_017006805.1 105 UTR 5 XP_016862294.1
XM_017006806.1 105 UTR 5 XP_016862295.1
XM_017006807.1 105 Silent Mutation CAA,CAG Q23Q XP_016862296.1

View Full Product Details