Product Details

SNP ID
rs115866195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:55468531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTCAAAAAACTTTTGGAATAAT[C/G]CCAACTGGACTGCAGTGAAGGACAA
Phenotype
MIM: 164975
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WNT5A PubMed Links

Gene Details

Gene
WNT5A
Gene Name
Wnt family member 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256105.1 2913 UTR 3 NP_001243034.1
NM_003392.4 2913 UTR 3 NP_003383.2
XM_006713324.1 2913 UTR 3 XP_006713387.1
XM_011534085.2 2913 UTR 3 XP_011532387.1
XM_011534086.2 2913 UTR 3 XP_011532388.1
XM_011534087.2 2913 UTR 3 XP_011532389.1
XM_011534088.2 2913 UTR 3 XP_011532390.1
XM_011534089.1 2913 UTR 3 XP_011532391.1
XM_017007127.1 2913 UTR 3 XP_016862616.1
XM_017007128.1 2913 UTR 3 XP_016862617.1

View Full Product Details