Product Details

SNP ID
rs117956843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9937549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCATGTTTCCCACCAGCCCTGC[A/C]CTGTCCGATCAGTGAGACCCGCCTG
Phenotype
MIM: 607170 MIM: 610925
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRELD1 PubMed Links

Gene Details

Gene
CRELD1
Gene Name
cysteine rich with EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031717.3 Intron NP_001026887.1
NM_001077415.2 Intron NP_001070883.1
NM_015513.4 Intron NP_056328.2
XM_011534108.1 Intron XP_011532410.1
XM_017007175.1 Intron XP_016862664.1
Gene
IL17RC
Gene Name
interleukin 17 receptor C
There are no transcripts associated with this gene.

Gene
PRRT3
Gene Name
proline rich transmembrane protein 3
There are no transcripts associated with this gene.

Gene
PRRT3-AS1
Gene Name
PRRT3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details