Product Details
- SNP ID
-
rs116978549
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70055524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGCATCATTCACATCGAGAATT[G/T]CCATTTTATGGGGACTATGGATCAA
- Phenotype
-
MIM: 142701
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HTN1
PubMed Links
Gene Details
- Gene
- HTN1
- Gene Name
- histatin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002159.3 |
223 |
Missense Mutation |
TTG,TTT |
L43F |
NP_002150.1 |
View Full Product Details