Product Details

SNP ID
rs114626462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70816941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATATACTGACAGAAAATGATCTT[C/T]AGGGTGTTTAGGTGGGAGAAAAGCC
Phenotype
MIM: 604851 MIM: 611194
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRSF1 PubMed Links

Gene Details

Gene
GRSF1
Gene Name
G-rich RNA sequence binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098477.1 5308 UTR 3 NP_001091947.1
NM_002092.3 5308 UTR 3 NP_002083.3
XM_005265681.3 5308 Intron XP_005265738.1
XM_011531897.2 5308 Intron XP_011530199.1
Gene
RUFY3
Gene Name
RUN and FYVE domain containing 3
There are no transcripts associated with this gene.

View Full Product Details