Product Details

SNP ID
rs114386213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55079436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCAAAGCAAGGTAACGTTAGT[C/T]TTCCTTTCTATCAATCCGAATTCCA
Phenotype
MIM: 191306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDR PubMed Links

Gene Details

Gene
KDR
Gene Name
kinase insert domain receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002253.2 4878 UTR 3 NP_002244.1

View Full Product Details