Product Details

SNP ID
rs111731693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55855851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATAGTGTCTGGGCCCTACAACA[C/T]ATTAAGGGATTAGCAGCATGGACAC
Phenotype
MIM: 607879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC1 PubMed Links

Gene Details

Gene
EXOC1
Gene Name
exocyst complex component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024924.1 Intron NP_001020095.1
NM_018261.3 Intron NP_060731.2
NM_178237.2 Intron NP_839955.1
XM_005265747.2 Intron XP_005265804.1
XM_005265748.1 Intron XP_005265805.1
XM_005265750.2 Intron XP_005265807.1
XM_017008405.1 Intron XP_016863894.1
XM_017008406.1 Intron XP_016863895.1
XM_017008407.1 Intron XP_016863896.1
XM_017008408.1 Intron XP_016863897.1
XM_017008409.1 Intron XP_016863898.1

View Full Product Details