Product Details

SNP ID
rs113304533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177403300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCACCTGGCAGAGCGTGGTCTC[C/G]GAGGGTCGCGCGGCGCCGCTTGGCA
Phenotype
MIM: 610619 MIM: 612812 MIM: 182309
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
F12 PubMed Links

Gene Details

Gene
F12
Gene Name
coagulation factor XII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000505.3 1534 Silent Mutation TCC,TCG S495S NP_000496.2
XM_011534462.2 1534 Silent Mutation TCC,TCG S383S XP_011532764.1
Gene
PFN3
Gene Name
profilin 3
There are no transcripts associated with this gene.

Gene
SLC34A1
Gene Name
solute carrier family 34 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167579.1 1534 Intron NP_001161051.1
NM_003052.4 1534 Intron NP_003043.3
XM_005265975.1 1534 Intron XP_005266032.1
XM_017009773.1 1534 Intron XP_016865262.1
XM_017009774.1 1534 Intron XP_016865263.1
XM_017009775.1 1534 Intron XP_016865264.1

View Full Product Details