Product Details

SNP ID
rs80358597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32316494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATCCAAAGAGAGGCCAACATTT[G/T]TTGAAATTTTTAAGACACGCTGCAA
Phenotype
MIM: 600185
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BRCA2 PubMed Links

Gene Details

Gene
BRCA2
Gene Name
BRCA2, DNA repair associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000059.3 261 Missense Mutation GTT,TTT V12F NP_000050.2
Gene
ZAR1L
Gene Name
zygote arrest 1-like
There are no transcripts associated with this gene.

View Full Product Details