Product Details
- SNP ID
-
rs117592751
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:42888078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATTTGTACCATAAAGAAAGTAGG[C/G]ATTAAAATCTAAAAAGACCCCCAAA
- Phenotype
-
MIM: 607441
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EPSTI1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11385952] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPSTI1
- Gene Name
- epithelial stromal interaction 1 (breast)
View Full Product Details