Product Details

SNP ID
rs114163392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:75285762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAAAAGAAAAAATGTATTTCAA[A/C]ACAGCTTGTCATCTTTGGAAAAAAT
Phenotype
MIM: 612465
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TBC1D4 PubMed Links

Gene Details

Gene
TBC1D4
Gene Name
TBC1 domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286658.2 5328 UTR 3 NP_001273587.1
NM_001286659.2 5328 UTR 3 NP_001273588.1
NM_014832.4 5328 UTR 3 NP_055647.2
XM_005266603.1 5328 UTR 3 XP_005266660.1
XM_005266605.2 5328 UTR 3 XP_005266662.1
XM_006719903.3 5328 UTR 3 XP_006719966.1
XM_011535331.1 5328 UTR 3 XP_011533633.1
XM_017020882.1 5328 UTR 3 XP_016876371.1
XM_017020883.1 5328 UTR 3 XP_016876372.1
XM_017020884.1 5328 UTR 3 XP_016876373.1

View Full Product Details