Product Details

SNP ID
rs117425231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:77045048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTTGCTTGAAAGCCAGCAT[C/T]GTTCTTAGTCCATGGGCATGGCGAT
Phenotype
MIM: 610392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCBP2 PubMed Links

Gene Details

Gene
MYCBP2
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015057.4 14502 UTR 3 NP_055872.4
XM_005266299.2 14502 UTR 3 XP_005266356.1
XM_006719779.2 14502 UTR 3 XP_006719842.1
XM_006719780.2 14502 UTR 3 XP_006719843.1
XM_006719781.2 14502 UTR 3 XP_006719844.1
XM_006719782.2 14502 UTR 3 XP_006719845.1
XM_006719783.2 14502 UTR 3 XP_006719846.1
XM_006719784.2 14502 UTR 3 XP_006719847.1
XM_006719785.2 14502 UTR 3 XP_006719848.1
XM_006719786.2 14502 UTR 3 XP_006719849.1
XM_006719787.2 14502 UTR 3 XP_006719850.1
XM_006719788.2 14502 UTR 3 XP_006719851.1
XM_006719789.2 14502 UTR 3 XP_006719852.1
XM_011535004.2 14502 UTR 3 XP_011533306.1
XM_011535005.2 14502 UTR 3 XP_011533307.1
XM_017020454.1 14502 UTR 3 XP_016875943.1
XM_017020455.1 14502 UTR 3 XP_016875944.1
XM_017020456.1 14502 UTR 3 XP_016875945.1
XM_017020457.1 14502 UTR 3 XP_016875946.1
XM_017020458.1 14502 UTR 3 XP_016875947.1
XM_017020459.1 14502 UTR 3 XP_016875948.1
XM_017020460.1 14502 UTR 3 XP_016875949.1
XM_017020461.1 14502 UTR 3 XP_016875950.1
XM_017020462.1 14502 UTR 3 XP_016875951.1
XM_017020463.1 14502 UTR 3 XP_016875952.1
XM_017020464.1 14502 UTR 3 XP_016875953.1
XM_017020465.1 14502 UTR 3 XP_016875954.1
XM_017020466.1 14502 UTR 3 XP_016875955.1
XM_017020467.1 14502 UTR 3 XP_016875956.1
XM_017020468.1 14502 UTR 3 XP_016875957.1

View Full Product Details