Product Details

SNP ID
rs115210819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28300428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATATAGGACCAAACCATGTCT[C/G]TCTTATATCTGTAGCATATATTCTT
Phenotype
MIM: 165070
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLT1 PubMed Links

Gene Details

Gene
FLT1
Gene Name
fms related tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159920.1 7041 Intron NP_001153392.1
NM_001160030.1 7041 Intron NP_001153502.1
NM_001160031.1 7041 Intron NP_001153503.1
NM_002019.4 7041 UTR 3 NP_002010.2
XM_011535014.1 7041 Intron XP_011533316.1
XM_017020485.1 7041 UTR 3 XP_016875974.1
Gene
PAN3
Gene Name
PAN3 poly(A) specific ribonuclease subunit
There are no transcripts associated with this gene.

View Full Product Details