Product Details

SNP ID
rs116790577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79069437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGACCAGATCCCGCTTTACCC[G/T]GTGCGTAGCGCAGCGGCGGCCGCAG
Phenotype
MIM: 614446
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP9B PubMed Links

Gene Details

Gene
ATP9B
Gene Name
ATPase phospholipid transporting 9B (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306085.1 163 Silent Mutation CCG,CCT P9P NP_001293014.1
NM_198531.4 163 Silent Mutation CCG,CCT P9P NP_940933.3
XM_011525963.2 163 UTR 5 XP_011524265.1
XM_011525964.2 163 Silent Mutation CCG,CCT P9P XP_011524266.1
XM_011525965.2 163 UTR 5 XP_011524267.1
XM_011525966.2 163 Intron XP_011524268.1
XM_011525971.2 163 Intron XP_011524273.2
XM_011525972.2 163 Intron XP_011524274.1
XM_011525973.2 163 Intron XP_011524275.1
XM_011525974.2 163 Intron XP_011524276.1
XM_017025726.1 163 UTR 5 XP_016881215.1
XM_017025727.1 163 UTR 5 XP_016881216.1
XM_017025728.1 163 UTR 5 XP_016881217.1
XM_017025729.1 163 UTR 5 XP_016881218.1
XM_017025730.1 163 UTR 5 XP_016881219.1
XM_017025731.1 163 UTR 5 XP_016881220.1
XM_017025732.1 163 UTR 5 XP_016881221.1
XM_017025733.1 163 UTR 5 XP_016881222.1
XM_017025734.1 163 UTR 5 XP_016881223.1
XM_017025735.1 163 UTR 5 XP_016881224.1
XM_017025736.1 163 UTR 5 XP_016881225.1
XM_017025737.1 163 UTR 5 XP_016881226.1
XM_017025738.1 163 Intron XP_016881227.1
XM_017025739.1 163 Intron XP_016881228.1
XM_017025740.1 163 Intron XP_016881229.1
XM_017025741.1 163 Intron XP_016881230.1
XM_017025742.1 163 UTR 5 XP_016881231.1

View Full Product Details