Product Details

SNP ID
rs111669307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57601775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATCCTCCATGGGGACATGTACC[A/G]TATTTTCTCTGTTTAAAAAAAGAAA
Phenotype
MIM: 108410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NARS PubMed Links

Gene Details

Gene
NARS
Gene Name
asparaginyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004539.3 1752 Silent Mutation TAC,TAT Y508Y NP_004530.1
XM_005266700.1 1752 Silent Mutation TAC,TAT Y507Y XP_005266757.1

View Full Product Details