Product Details

SNP ID
rs111342680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62523917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCGGGGCCGCTGGCCGACTTC[C/T]GAGAGCCCGCGGTGCGCTCGCGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC2 PubMed Links

Gene Details

Gene
ZCCHC2
Gene Name
zinc finger CCHC-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017742.5 270 Nonsense Mutation CGA,TGA R165* NP_060212.4
XM_006722493.3 270 Nonsense Mutation CGA,TGA R85* XP_006722556.1
XM_011526044.1 270 Intron XP_011524346.1
XM_017025802.1 270 Nonsense Mutation CGA,TGA R85* XP_016881291.1

View Full Product Details