Product Details

SNP ID
rs111491614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62524299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGAGGCTCCGCGCCGCGCTCC[C/G]CGGGGGCCCCGAGGACGCGGAGGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZCCHC2 PubMed Links

Gene Details

Gene
ZCCHC2
Gene Name
zinc finger CCHC-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017742.5 652 Missense Mutation CCC,CGC P292R NP_060212.4
XM_006722493.3 652 Missense Mutation CCC,CGC P212R XP_006722556.1
XM_011526044.1 652 Intron XP_011524346.1
XM_017025802.1 652 Missense Mutation CCC,CGC P212R XP_016881291.1

View Full Product Details